Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318391.1 | NP_001305320.1 | 2 | 122 | ||
NM_032830.2 | NP_116219.2 | 2 | 176 | |||
GenBank mRNA | AB075868.2 | - | 2 | 114 | ||
AK027634.1 | - | 2 | 128 | |||
AK027675.1 | - | 2 | 125 | |||
AK074795.1 | - | 2 | 142 | |||
AK310104.1 | - | 2 | 69 | |||
BC009348.2 | - | 2 | 108 | 4131625 | ||
BX647265.1 | - | 2 | 90 | |||
DA277545.1 | - | 2 | 122 | |||
DC368305.1 | - | 2 | 176 |