Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013620.3 | NP_001013642.1 | 3 | 1292 | ||
XM_005268665.4 | XP_005268722.1 | 3 | 1073 | |||
GenBank mRNA | AY845858.1 | - | 3 | 1153 | ||
BC137413.1 | - | 3 | 1208 | 9021411 | ||
BC137414.1 | - | 3 | 1208 | 9021412 | ||
BC144617.1 | - | 3 | 1182 | |||
DB224774.1 | - | 1 | 135 |
Gene Symbol: ALG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032834.3 | NP_116223.3 | 3 | 1472 | ||
GenBank mRNA | AJ312278.1 | - | 3 | 1472 | ||
AK292488.1 | - | 3 | 1238 | |||
BC033730.1 | - | 3 | 1253 | |||
BC070347.1 | - | 3 | 1257 | 3839674 | ||
BC090948.1 | - | 3 | 1239 | 5205402 |