Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077268.1 | NP_001070736.1 | 1, 2 | 798 | ||
NM_001258420.1 | NP_001245349.1 | 1, 2 | 798 | |||
XM_017022684.1 | XP_016878173.1 | 1, 2 | 785 | |||
GenBank mRNA | AK027746.1 | - | 1, 2 | 230 | ||
AK225553.1 | - | 1, 2 | 221 | |||
BC021092.1 | - | 1, 2 | 95 | 4662224 | ||
DB012963.1 | - | 1, 2 | 216 |