Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077268.1 | NP_001070736.1 | 1 | 779 | ||
NM_001258420.1 | NP_001245349.1 | 1 | 779 | |||
XM_017022683.1 | XP_016878172.1 | 1 | 174 | |||
XM_017022684.1 | XP_016878173.1 | 1 | 766 | |||
GenBank mRNA | AB209224.1 | - | 447 | |||
AK027746.1 | - | 1 | 211 | |||
AK225553.1 | - | 1 | 202 | |||
BC021092.1 | - | 1 | 76 | 4662224 | ||
DB012963.1 | - | 1 | 197 |