Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032268.4 | NP_115644.1 | 4 | 1297 | ||
XM_017023793.1 | XP_016879282.1 | 4 | 1271 | |||
GenBank mRNA | AF378524.1 | - | 4 | 969 | ||
AK094870.1 | - | 1 | 1700 | |||
AK128349.1 | - | 1 | 2438 | |||
AL136903.1 | - | 4 | 962 | |||
AL390144.1 | - | 1 | 1115 | |||
AL834440.1 | - | 5 | 774 | |||
BC007235.1 | - | 4 | 969 | 2823677 |