Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032853.3 | NP_116242.2 | 5 | 590 | ||
NR_024247.1 | 5 | 659 | ||||
XM_011528380.2 | XP_011526682.1 | 5 | 580 | |||
XM_011528382.2 | XP_011526684.1 | 5 | 580 | |||
XM_011528383.1 | XP_011526685.1 | 5 | 580 | |||
XM_011528384.2 | XP_011526686.1 | 5 | 580 | |||
XM_011528385.2 | XP_011526687.1 | 6 | 781 | |||
XM_017027401.1 | XP_016882890.1 | 5 | 580 | |||
GenBank mRNA | AK309092.1 | - | 4 | 478 | ||
AK310721.1 | - | 6 | 693 | |||
BC110874.1 | - | 4 | 556 | 4933735 | ||
BC130443.1 | - | 5 | 619 | 40146474 | ||
BC144138.1 | - | 4 | 506 | 9052658 | ||
BC144147.1 | - | 5 | 619 | |||
BQ895780.1 | - | 5 | 649 | |||
BU541563.1 | - | 5 | 629 | |||
U20896.1 | - | 3 | 1588 | |||
U20897.1 | - | 4 | 560 |