Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSH2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291274.1 | NP_001278203.1 | 8 | 2533 | ||
NM_032855.3 | NP_116244.1 | 8 | 2000 | |||
NR_111903.1 | 8 | 2007 | ||||
XM_011528386.2 | XP_011526688.1 | 7 | 1754 | |||
XM_011528387.2 | XP_011526689.1 | 7 | 1756 | |||
XM_011528388.2 | XP_011526690.1 | 8 | 1834 | |||
XM_011528389.2 | XP_011526691.1 | 6 | 1621 | |||
XM_017027402.1 | XP_016882891.1 | 6 | 3154 | |||
XM_017027403.1 | XP_016882892.1 | 7 | 1708 | |||
XM_017027404.1 | XP_016882893.1 | 6 | 1592 | |||
GenBank mRNA | AK027792.1 | - | 6 | 1580 | ||
AK131222.1 | - | 6 | 2113 | |||
AY319652.1 | - | 8 | 2000 | |||
BC016826.1 | - | 4 | 1205 | |||
BC025237.2 | - | 5 | 1500 | 4853087 |