Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135040.2 | NP_001128512.1 | 21 | 3109 | ||
NM_001135041.2 | NP_001128513.1 | 19 | 2832 | |||
NM_001190836.1 | NP_001177765.1 | 21 | 3052 | |||
NM_001190837.1 | NP_001177766.1 | 23 | 3148 | |||
NM_004082.4 | NP_004073.2 | 24 | 3169 | |||
NM_023019.3 | NP_075408.1 | 19 | 2832 | |||
NR_033935.1 | 20 | 3135 | ||||
GenBank mRNA | AB209625.1 | - | 14 | 3830 | ||
AK297286.1 | - | 21 | 2804 | |||
AK314352.1 | - | 21 | 3053 | |||
AK315014.1 | - | 19 | 2624 | |||
BC071583.1 | - | 19 | 2749 | 4817849 | ||
BX640799.1 | - | 24 | 2948 | |||
BX647925.1 | - | 20 | 2896 | |||
BX647972.1 | - | 20 | 2915 | |||
X98801.1 | - | 22 | 2806 |