Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPND | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300862.1 | NP_001287791.1 | 7 | 944 | ||
NM_032868.5 | NP_116257.2 | 7 | 944 | |||
XM_006722926.2 | XP_006722989.1 | 7 | 905 | |||
GenBank mRNA | AK308468.1 | - | 7 | 905 | ||
BC032652.1 | - | 7 | 901 | 5503607 | ||
BG696748.1 | - | 5 | 530 | |||
CR977937.1 | - | 4 | 281 |