Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135040.2 | NP_001128512.1 | 26 | 3819 | ||
NM_001135041.2 | NP_001128513.1 | 24 | 3542 | |||
NM_001190836.1 | NP_001177765.1 | 26 | 3762 | |||
NM_001190837.1 | NP_001177766.1 | 29 | 3873 | |||
NM_004082.4 | NP_004073.2 | 30 | 3894 | |||
NM_023019.3 | NP_075408.1 | 25 | 3557 | |||
NR_033935.1 | 25 | 3845 | ||||
GenBank mRNA | AB209625.1 | - | 18 | 4814 | ||
AK055298.1 | - | 3 | 1839 | |||
AK297286.1 | - | 26 | 3514 | |||
AK314352.1 | - | 26 | 3763 | |||
AK315014.1 | - | 24 | 3334 | |||
BC006163.1 | - | 3 | 339 | |||
BC071583.1 | - | 24 | 3459 | 4817849 | ||
BT006758.1 | - | 3 | 336 | |||
BX640799.1 | - | 31 | 3673 | |||
BX647925.1 | - | 25 | 3606 | |||
BX647972.1 | - | 25 | 3625 | |||
X98801.1 | - | 28 | 3531 |