Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032871.3 | NP_116260.2 | 11 | 2174 | ||
NM_152222.1 | NP_689408.1 | 11 | 2112 | |||
XM_005274356.2 | XP_005274413.1 | 6 | 2275 | |||
XM_006718714.2 | XP_006718777.1 | 9 | 1739 | |||
XM_011545305.1 | XP_011543607.1 | 10 | 1997 | |||
GenBank mRNA | AK027899.1 | - | 11 | 2112 | ||
AK057036.1 | - | 5 | 1916 | |||
AK074128.1 | - | 3 | 3847 | |||
BC017279.2 | - | 11 | 2037 | 4873951 | ||
BC051810.1 | - | 11 | 2063 | 5216988 |