Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBASH3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032873.4 | NP_116262.2 | 12 | 2023 | ||
XM_005271712.3 | XP_005271769.1 | 12 | 4269 | |||
XM_011543041.2 | XP_011541343.1 | 12 | 1661 | |||
XM_011543042.1 | XP_011541344.1 | 12 | 2027 | |||
GenBank mRNA | AB075839.1 | - | 12 | 1875 | ||
AF425252.1 | - | 10 | 1320 | |||
AK075203.1 | - | 12 | 1991 | |||
AK222843.1 | - | 12 | 1973 | |||
AK222846.1 | - | 12 | 1973 | |||
BC007541.1 | - | 12 | 1977 | 2958242 |