Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170790.1 | NP_001164261.1 | 2 | 595 | ||
NM_032889.4 | NP_116278.3 | 2 | 322 | |||
XM_005269197.1 | XP_005269254.1 | 3 | 850 | |||
XM_005269198.4 | XP_005269255.1 | 2 | 272 | |||
GenBank mRNA | AK074684.1 | - | 2 | 319 | ||
AK097576.1 | - | 2 | 528 | |||
AK225998.1 | - | 2 | 207 | |||
AY358604.1 | - | 2 | 251 | |||
BC007703.2 | - | 1 | 138 | 3948896 | ||
BC067795.1 | - | 2 | 250 | 5299341 | ||
BX640649.1 | - | 2 | 216 | |||
DA428788.1 | - | 2 | 322 | |||
DA695979.1 | - | 2 | 321 |