Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032890.3 | NP_116279.2 | 10 | 4528 | ||
XM_005273335.2 | XP_005273392.1 | 8 | 4369 | |||
XM_006711592.2 | XP_006711655.1 | 9 | 5316 | |||
XM_006711594.3 | XP_006711657.1 | 8 | 4024 | |||
XM_011510072.2 | XP_011508374.1 | 9 | 4368 | |||
XM_011510073.2 | XP_011508375.1 | 9 | 4846 | |||
XM_011510074.2 | XP_011508376.1 | 9 | 4387 | |||
XM_011510075.2 | XP_011508377.1 | 9 | 4874 | |||
XM_011510077.2 | XP_011508379.1 | 8 | 4310 | |||
XM_017002611.1 | XP_016858100.1 | 8 | 4784 | |||
XM_017002612.1 | XP_016858101.1 | 9 | 4193 | |||
XM_017002613.1 | XP_016858102.1 | 7 | 4153 | |||
GenBank mRNA | AK023679.1 | - | 1 | 1855 | ||
AK026114.1 | - | 1 | 2418 | |||
AK056569.1 | - | 3 | 3332 | |||
AL133092.1 | - | 1 | 1902 | |||
BC011542.1 | - | 1 | 2663 |