Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204300.1 | NP_001191229.1 | 3 | 432 | ||
NM_001256423.1 | NP_001243352.1 | 3 | 415 | |||
NM_032900.5 | NP_116289.4 | 3 | 432 | |||
GenBank mRNA | AK090447.1 | - | 3 | 382 | ||
AK093441.1 | - | 3 | 389 | |||
AK303055.1 | - | 3 | 393 | |||
BC113888.1 | - | 3 | 408 | 40082328 | ||
BC114490.1 | - | 3 | 408 | 40082327 | ||
BI830638.1 | - | 3 | 417 | |||
BX648189.1 | - | 3 | 407 | |||
CR749412.1 | - | 3 | 408 | |||
DB069205.1 | - | 3 | 415 | |||
DQ338460.1 | - | 3 | 397 |
Gene Symbol: ARHGAP19-SLIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037909.1 | 3 | 432 | |||
GenBank mRNA | AY336750.1 | - | 3 | 397 |