Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005272350.2 | XP_005272407.1 | 3 | 1127 | ||
XM_011517349.2 | XP_011515651.1 | 1 | 94 | |||
XM_017013912.1 | XP_016869401.1 | 4 | 1196 | |||
XM_017013913.1 | XP_016869402.1 | 3 | 1160 | |||
XM_017013914.1 | XP_016869403.1 | 4 | 1234 | |||
XM_017013915.1 | XP_016869404.1 | 3 | 1191 | |||
GenBank mRNA | AK090471.1 | - | 3 | 1057 | ||
BC007854.2 | - | 1 | 201 | 4298333 | ||
BC049841.1 | - | 1 | 453 | 4991947 | ||
BC053506.1 | - | 1 | 542 | 2823232 | ||
BT006845.1 | - | 1 | 73 |