Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005272350.2 | XP_005272407.1 | 12 | 2325 | ||
XM_011517349.2 | XP_011515651.1 | 11 | 1176 | |||
XM_017013912.1 | XP_016869401.1 | 13 | 2394 | |||
XM_017013913.1 | XP_016869402.1 | 12 | 2358 | |||
XM_017013914.1 | XP_016869403.1 | 13 | 2432 | |||
XM_017013915.1 | XP_016869404.1 | 12 | 2389 | |||
GenBank mRNA | AK090471.1 | - | 9 | 3691 | ||
BC007854.2 | - | 10 | 1399 | 4298333 | ||
BC049841.1 | - | 10 | 1651 | 4991947 | ||
BC053506.1 | - | 10 | 1740 | 2823232 | ||
BT006845.1 | - | 10 | 1271 |