Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190726.1 | NP_001177655.1 | 4 | 600 | ||
NM_032918.2 | NP_116307.1 | 5 | 657 | |||
XM_017020121.1 | XP_016875610.1 | 6 | 1163 | |||
GenBank mRNA | AF339750.1 | - | 4 | 400 | ||
AK295343.1 | - | 4 | 540 | |||
AK311016.1 | - | 4 | 326 | |||
AK313881.1 | - | 5 | 597 | |||
BC007997.1 | - | 5 | 550 | 3357341 |