Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYKPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278346.1 | NP_001265275.1 | 4 | 1495 | ||
NM_153373.3 | NP_699204.1 | 4 | 672 | |||
NR_103509.1 | 3 | 1376 | ||||
NR_103510.1 | 6 | 893 | ||||
XM_006714925.2 | XP_006714988.1 | 4 | 634 | |||
XM_011534669.2 | XP_011532971.1 | 4 | 634 | |||
XM_011534670.2 | XP_011532972.1 | 3 | 967 | |||
XM_011534671.2 | XP_011532973.1 | 4 | 634 | |||
XM_011534672.2 | XP_011532974.1 | 4 | 420 | |||
XM_011534676.2 | XP_011532978.1 | 1 | 662 | |||
XM_017009995.1 | XP_016865484.1 | 4 | 634 | |||
XM_017009996.1 | XP_016865485.1 | 4 | 634 | |||
XM_017009997.1 | XP_016865486.1 | 4 | 634 | |||
XM_017009998.1 | XP_016865487.1 | 4 | 1092 | |||
GenBank mRNA | AK023470.1 | - | 1 | 712 | ||
AK292061.1 | - | 4 | 634 | |||
BC028002.1 | - | 3 | 1327 | |||
BC037567.1 | - | 4 | 634 | 5220434 | ||
BC067745.1 | - | 3 | 343 | |||
BC110335.1 | - | 4 | 644 | 5753177 | ||
BX417194.2 | - | 4 | 724 |