Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297551.1 | NP_001284480.1 | 2 | 224 | ||
NM_001297552.1 | NP_001284481.1 | 2 | 224 | |||
NM_032927.3 | NP_116316.1 | 2 | 729 | |||
XM_005248034.2 | XP_005248091.1 | 2 | 247 | |||
GenBank mRNA | AK295248.1 | - | 2 | 206 | ||
AK296007.1 | - | 2 | 205 | |||
BC007729.2 | - | 2 | 694 | 4303698 | ||
BC091482.1 | - | 2 | 224 | 30344390 | ||
BM263799.1 | - | 2 | 206 | |||
CR933611.1 | - | 2 | 214 | |||
DA019506.1 | - | 2 | 214 |