Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032888.3 | NP_116277.2 | 59 | 5525 | ||
XM_006717308.2 | XP_006717371.1 | 58 | 5539 | |||
XM_006717310.3 | XP_006717373.1 | 57 | 3338 | |||
XM_011519138.2 | XP_011517440.1 | 59 | 5157 | |||
XM_011519140.1 | XP_011517442.1 | 58 | 5539 | |||
XM_011519145.2 | XP_011517447.1 | 46 | 2767 | |||
GenBank mRNA | AB058773.1 | - | 56 | 4012 | ||
AK021957.1 | - | 1 | 270 | |||
AY149237.1 | - | 59 | 5119 |