Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032888.3 | NP_116277.2 | 3 | 1064 | ||
XM_006717308.2 | XP_006717371.1 | 3 | 1132 | |||
XM_011519138.2 | XP_011517440.1 | 3 | 696 | |||
XM_011519140.1 | XP_011517442.1 | 3 | 1132 | |||
XM_011519142.2 | XP_011517444.1 | 3 | 1138 | |||
XM_011519143.2 | XP_011517445.1 | 3 | 1136 | |||
XM_011519144.2 | XP_011517446.1 | 3 | 1136 | |||
XM_017015239.1 | XP_016870728.1 | 3 | 1137 | |||
GenBank mRNA | AY149237.1 | - | 3 | 658 |