Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102450.2 | NP_001095920.1 | 4, 5 | 413 | ||
NM_033345.3 | NP_203131.1 | 3, 4 | 284 | |||
XM_005245555.3 | XP_005245612.1 | 4, 5 | 427 | |||
XM_011510089.2 | XP_011508391.1 | 4, 5 | 1324 | |||
XM_011510090.2 | XP_011508392.1 | 7, 8 | 1154 | |||
XM_011510091.2 | XP_011508393.1 | 3, 4 | 301 | |||
XM_017002631.1 | XP_016858120.1 | 7, 8 | 774 | |||
XM_017002632.1 | XP_016858121.1 | 5, 6 | 605 | |||
XM_017002633.1 | XP_016858122.1 | 4, 5 | 803 | |||
XM_017002634.1 | XP_016858123.1 | 4, 5 | 482 | |||
XM_017002635.1 | XP_016858124.1 | 5, 6 | 929 | |||
XM_017002636.1 | XP_016858125.1 | 3, 4 | 411 | |||
XM_017002637.1 | XP_016858126.1 | 4, 5 | 609 | |||
GenBank mRNA | AF300649.1 | - | 2, 3 | 122 | ||
AK294660.1 | - | 4, 5 | 380 | |||
AK296514.1 | - | 4, 5 | 373 | |||
BC069677.1 | - | 4, 5 | 362 | 7262579 | ||
BC069718.1 | - | 3, 4 | 234 | 7262591 | ||
BC103865.1 | - | 4, 5 | 389 | 40003386 | ||
BC103866.1 | - | 4, 5 | 389 | 40003387 | ||
BC103867.1 | - | 4, 5 | 389 | 40003388 | ||
BG195915.1 | - | 4, 5 | 414 | |||
BX281622.1 | - | 3, 4 | 295 | |||
BX954018.1 | - | 3, 4 | 188 |