Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033083.6 | NP_149074.3 | 3 | 465 | ||
GenBank mRNA | AF272973.1 | - | 3 | 396 | ||
AK312330.1 | - | 3 | 437 | |||
AW292089.1 | - | 3 | 170 | |||
BC041329.2 | - | 3 | 407 | 5284115 |