Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033083.6 | NP_149074.3 | 5 | 770 | ||
XM_005265518.1 | XP_005265575.1 | 4 | 633 | |||
XM_011534165.1 | XP_011532467.1 | 4 | 782 | |||
XM_011534166.1 | XP_011532468.1 | 4 | 777 | |||
GenBank mRNA | AF272973.1 | - | 5 | 701 | ||
AK304697.1 | - | 4 | 592 | |||
AK312330.1 | - | 5 | 742 | |||
AW292089.1 | - | 5 | 475 | |||
BC041329.2 | - | 5 | 712 | 5284115 | ||
BQ064584.1 | - | 2 | 73 |