Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033119.4 | NP_149110.1 | 9 | 984 | ||
XM_011523399.2 | XP_011521701.1 | 12 | 1295 | |||
XM_011523400.2 | XP_011521702.1 | 7 | 586 | |||
XM_017023804.1 | XP_016879293.1 | 11 | 2514 | |||
GenBank mRNA | AB062886.1 | - | 9 | 765 | ||
AF289584.1 | - | 9 | 1036 | |||
AF358135.1 | - | 9 | 767 | |||
AK314930.1 | - | 9 | 918 | |||
AY061883.1 | - | 9 | 918 | |||
BC051288.1 | - | 9 | 984 | 6646089 |