Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033119.4 | NP_149110.1 | 10 | 1249 | ||
XM_011523399.2 | XP_011521701.1 | 13 | 1560 | |||
XM_011523400.2 | XP_011521702.1 | 8 | 851 | |||
XM_017023804.1 | XP_016879293.1 | 12 | 2779 | |||
GenBank mRNA | AB062886.1 | - | 10 | 1030 | ||
AF289584.1 | - | 10 | 1301 | |||
AF358135.1 | - | 10 | 1032 | |||
AK314930.1 | - | 10 | 1183 | |||
AY061883.1 | - | 10 | 1183 | |||
BC051288.1 | - | 10 | 1249 | 6646089 |