Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033125.3 | NP_149116.2 | 4 | 1107 | ||
XM_005267184.3 | XP_005267241.1 | 4 | 756 | |||
XM_011536204.2 | XP_011534506.1 | 4 | 1121 | |||
XM_011536205.2 | XP_011534507.1 | 4 | 1079 | |||
XM_011536206.2 | XP_011534508.1 | 5 | 809 | |||
XM_011536207.2 | XP_011534509.1 | 4 | 816 | |||
XM_011536208.2 | XP_011534510.1 | 4 | 729 | |||
XM_011536209.2 | XP_011534511.1 | 5 | 871 | |||
XM_011536210.1 | XP_011534512.1 | 4 | 1079 | |||
XM_011536211.2 | XP_011534513.1 | 3 | 611 | |||
XM_011536212.2 | XP_011534514.1 | 2 | 462 | |||
GenBank mRNA | AB055798.1 | - | 6 | 1222 | ||
AF268892.1 | - | 4 | 1040 | |||
AK057306.1 | - | 3 | 791 | |||
AY145502.1 | - | 4 | 1040 | |||
BC037247.1 | - | 4 | 1055 | |||
BC047565.1 | - | 4 | 1107 | 4838011 | ||
GQ472228.1 | - | 4 | 1107 |