Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033103.4 | NP_149094.3 | 15 | 1891 | ||
GenBank mRNA | AF268032.1 | - | 15 | 1849 | ||
AF423421.1 | - | 15 | 1825 | |||
AJ347750.1 | - | 15 | 1825 | |||
AK095001.1 | - | 6 | 661 | |||
AK098246.1 | - | 15 | 1861 | |||
AK126506.1 | - | 12 | 2159 | |||
AK300775.1 | - | 14 | 1785 | |||
AK315105.1 | - | 15 | 1861 | |||
AL831950.1 | - | 14 | 1739 | |||
BC036447.2 | - | 15 | 1861 | 4830913 |