Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033031.2 | NP_149020.2 | 11 | 4160 | ||
NM_033670.2 | NP_391990.1 | 7 | 848 | |||
XM_017029913.1 | XP_016885402.1 | 13 | 4371 | |||
XM_017029914.1 | XP_016885403.1 | 13 | 4301 | |||
XM_017029915.1 | XP_016885404.1 | 14 | 4331 | |||
XM_017029916.1 | XP_016885405.1 | 13 | 4368 | |||
XM_017029917.1 | XP_016885406.1 | 12 | 4347 | |||
XM_017029918.1 | XP_016885407.1 | 12 | 4344 | |||
GenBank mRNA | AJ314764.1 | - | 11 | 4164 | ||
AJ314765.1 | - | 7 | 852 | |||
AJ314766.1 | - | 10 | 1172 | |||
AJ416458.1 | - | 11 | 4176 | |||
AK292454.1 | - | 12 | 4333 | |||
AL137550.1 | - | 3 | 302 | |||
AY927611.1 | - | 4 | 1025 |