Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033114.3 | NP_149105.3 | 4 | 395 | ||
XM_017020124.1 | XP_016875613.1 | 3 | 286 | |||
GenBank mRNA | AB062361.1 | - | 4 | 395 | ||
AK308244.1 | - | 4 | 265 | |||
BC005099.1 | - | 4 | 345 | |||
BC010177.2 | - | 4 | 360 | |||
BC022543.1 | - | 4 | 370 | 4797962 |