Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033402.4 | NP_208325.3 | 11 | 1907 | ||
XM_005251318.3 | XP_005251375.1 | 10 | 1696 | |||
XM_005251319.4 | XP_005251376.1 | 9 | 1390 | |||
XM_005251320.3 | XP_005251377.1 | 8 | 1182 | |||
XM_005251322.3 | XP_005251379.1 | 7 | 996 | |||
XM_017013920.1 | XP_016869409.1 | 12 | 1855 | |||
XM_017013921.1 | XP_016869410.1 | 12 | 1855 | |||
XM_017013922.1 | XP_016869411.1 | 12 | 1855 | |||
XM_017013923.1 | XP_016869412.1 | 11 | 1682 | |||
XM_017013924.1 | XP_016869413.1 | 12 | 1855 | |||
XM_017013925.1 | XP_016869414.1 | 9 | 1401 | |||
XM_017013926.1 | XP_016869415.1 | 9 | 1384 | |||
XM_017013927.1 | XP_016869416.1 | 10 | 1412 | |||
XM_017013928.1 | XP_016869417.1 | 7 | 864 | |||
GenBank mRNA | AB051551.1 | - | 10 | 2528 | ||
AK056185.1 | - | 10 | 1870 | |||
AK057990.1 | - | 2 | 162 | |||
AK300234.1 | - | 8 | 1139 | |||
AK300874.1 | - | 10 | 1551 | |||
AK302648.1 | - | 11 | 1757 | |||
AK307350.1 | - | 6 | 766 | |||
AK307443.1 | - | 8 | 1163 | |||
AK307636.1 | - | 10 | 1541 | |||
BC030701.2 | - | 11 | 1780 | 4837403 |