Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080419.2 | NP_001073888.2 | 3 | 443 | ||
XM_006722145.3 | XP_006722208.1 | 6 | 4824 | |||
XM_006722148.2 | XP_006722211.1 | 4 | 512 | |||
XM_011525400.1 | XP_011523702.1 | 4 | 512 | |||
XM_017025247.1 | XP_016880736.1 | 6 | 4824 | |||
XM_017025248.1 | XP_016880737.1 | 5 | 4726 | |||
XM_017025249.1 | XP_016880738.1 | 5 | 869 | |||
XM_017025250.1 | XP_016880739.1 | 4 | 771 | |||
GenBank mRNA | AB051540.1 | - | 3 | 426 | ||
BC053362.1 | - | 3 | 426 |