Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282129.1 | NP_001269058.1 | 16 | 4401 | ||
NM_033389.3 | NP_203747.2 | 15 | 4132 | |||
XM_005258058.3 | XP_005258115.1 | 17 | 4294 | |||
XM_005258059.2 | XP_005258116.1 | 14 | 4268 | |||
XM_005258060.3 | XP_005258117.1 | 3 | 2861 | |||
XM_006722149.3 | XP_006722212.1 | 14 | 7899 | |||
XM_011525402.2 | XP_011523704.1 | 16 | 4129 | |||
XM_011525404.2 | XP_011523706.1 | 14 | 4892 | |||
XM_011525405.2 | XP_011523707.1 | 14 | 4940 | |||
XM_011525406.2 | XP_011523708.1 | 13 | 4219 | |||
XM_011525407.1 | XP_011523709.1 | 13 | 3964 | |||
XM_017025251.1 | XP_016880740.1 | 16 | 4337 | |||
XM_017025252.1 | XP_016880741.1 | 14 | 4198 | |||
GenBank mRNA | AB051512.1 | - | 3 | 2950 | ||
AB099290.1 | - | 15 | 4092 | |||
AK091432.1 | - | 2 | 2521 | |||
AK296413.1 | - | 16 | 4266 |