Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282129.1 | NP_001269058.1 | 13 | 1376 | ||
NM_033389.3 | NP_203747.2 | 12 | 1107 | |||
XM_005258058.3 | XP_005258115.1 | 14 | 1269 | |||
XM_005258059.2 | XP_005258116.1 | 11 | 1243 | |||
XM_006722149.3 | XP_006722212.1 | 11 | 4874 | |||
XM_011525402.2 | XP_011523704.1 | 13 | 1104 | |||
XM_011525404.2 | XP_011523706.1 | 11 | 1867 | |||
XM_011525405.2 | XP_011523707.1 | 11 | 1915 | |||
XM_011525406.2 | XP_011523708.1 | 10 | 1194 | |||
XM_011525407.1 | XP_011523709.1 | 10 | 939 | |||
XM_017025251.1 | XP_016880740.1 | 13 | 1312 | |||
XM_017025252.1 | XP_016880741.1 | 11 | 1173 | |||
XM_017025253.1 | XP_016880742.1 | 13 | 1315 | |||
XM_017025254.1 | XP_016880743.1 | 13 | 1315 | |||
GenBank mRNA | AB072358.1 | - | 12 | 1067 | ||
AB099290.1 | - | 12 | 1067 | |||
AF484838.1 | - | 3 | 226 | |||
AK296413.1 | - | 13 | 1241 |