Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282129.1 | NP_001269058.1 | 7 | 751 | ||
NM_001282130.1 | NP_001269059.1 | 6 | 670 | |||
NM_001282131.1 | NP_001269060.1 | 5 | 618 | |||
NM_033389.3 | NP_203747.2 | 6 | 482 | |||
XM_005258058.3 | XP_005258115.1 | 8 | 644 | |||
XM_005258059.2 | XP_005258116.1 | 5 | 618 | |||
XM_006722149.3 | XP_006722212.1 | 5 | 4249 | |||
XM_011525402.2 | XP_011523704.1 | 7 | 479 | |||
XM_011525404.2 | XP_011523706.1 | 5 | 1242 | |||
XM_011525405.2 | XP_011523707.1 | 5 | 1290 | |||
XM_011525406.2 | XP_011523708.1 | 4 | 569 | |||
XM_011525407.1 | XP_011523709.1 | 4 | 314 | |||
XM_017025251.1 | XP_016880740.1 | 7 | 687 | |||
XM_017025252.1 | XP_016880741.1 | 5 | 548 | |||
XM_017025253.1 | XP_016880742.1 | 7 | 690 | |||
XM_017025254.1 | XP_016880743.1 | 7 | 690 | |||
XM_017025255.1 | XP_016880744.1 | 7 | 690 | |||
GenBank mRNA | AB072358.1 | - | 6 | 442 | ||
AB072359.1 | - | 6 | 653 | |||
AB099290.1 | - | 6 | 442 | |||
AK296413.1 | - | 7 | 616 | |||
AK308887.1 | - | 6 | 549 | |||
BC008941.2 | - | 6 | 594 | |||
BC011636.2 | - | 5 | 576 | |||
BC068223.1 | - | 6 | 594 |