Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145546.1 | NP_001139018.1 | 3 | 228 | ||
NM_033106.3 | NP_149097.1 | 4 | 277 | |||
GenBank mRNA | AF188492.1 | - | 4 | 277 | ||
AF188493.1 | - | 4 | 277 |