Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033109.4 | NP_149100.2 | 9, 10 | 955 | ||
XM_005264629.2 | XP_005264686.1 | 9, 10 | 885 | |||
XM_017005172.1 | XP_016860661.1 | 8, 9 | 835 | |||
GenBank mRNA | AJ458465.1 | - | 9, 10 | 886 | ||
AK307589.1 | - | 885 | ||||
AY027528.1 | - | 9, 10 | 882 | |||
BC053660.1 | - | 9, 10 | 869 | 6062060 |