Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257191.2 | NP_001244120.1 | 10 | 1451 | ||
NM_004397.5 | NP_004388.2 | 10 | 1523 | |||
XM_005271417.3 | XP_005271474.1 | 10 | 1468 | |||
XM_011542644.1 | XP_011540946.1 | 9 | 1394 | |||
XM_011542645.1 | XP_011540947.1 | 9 | 1376 | |||
XM_017017251.1 | XP_016872740.1 | 9 | 1339 | |||
GenBank mRNA | AK093657.1 | - | 2 | 693 | ||
AK313244.1 | - | 10 | 1451 | |||
BC065007.1 | - | 9 | 1382 | 6163439 | ||
D17532.1 | - | 10 | 1396 | |||
Z11685.1 | - | 10 | 1423 |