Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HPS4 SIRNA Assays ›
Gene Symbol:
HPS4
Gene Name:
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases:
BLOC3S2, LE
Chromosome Location:
Chr.22: 26443613 - 26483863 on Build GRCh38
UniGene ID:
Hs.474436
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s40115
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HPS4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022081.5
NP_071364.4
5
994
NM_152841.2
NP_690054.1
3
611
NR_073135.1
5
903
NR_073136.1
3
611
XM_006724353.3
XP_006724416.1
5
985
XM_006724354.3
XP_006724417.1
5
890
XM_011530485.1
XP_011528787.1
5
985
XM_011530486.2
XP_011528788.1
5
890
XM_011530487.2
XP_011528789.1
6
1095
XM_011530488.2
XP_011528790.1
5
893
XM_011530489.2
XP_011528791.1
6
996
XM_011530490.2
XP_011528792.1
5
985
XM_011530491.2
XP_011528793.1
5
985
XM_011530492.1
XP_011528794.1
5
985
XM_011530493.2
XP_011528795.1
5
985
XM_011530494.2
XP_011528796.1
3
281
XM_017029042.1
XP_016884531.1
5
890
XM_017029043.1
XP_016884532.1
6
1095
XM_017029044.1
XP_016884533.1
5
893
XM_017029045.1
XP_016884534.1
5
985
XM_017029046.1
XP_016884535.1
5
985
XM_017029047.1
XP_016884536.1
5
985
XM_017029048.1
XP_016884537.1
5
985
XM_017029049.1
XP_016884538.1
5
890
XM_017029050.1
XP_016884539.1
5
985
XM_017029051.1
XP_016884540.1
5
890
XM_017029052.1
XP_016884541.1
5
985
XM_017029054.1
XP_016884543.1
5
985
XM_017029055.1
XP_016884544.1
4
894
XM_017029056.1
XP_016884545.1
5
985
XM_017029057.1
XP_016884546.1
5
985
XM_017029058.1
XP_016884547.1
5
868
XM_017029059.1
XP_016884548.1
5
894
XM_017029060.1
XP_016884549.1
5
893
XM_017029061.1
XP_016884550.1
5
985
XM_017029062.1
XP_016884551.1
4
894
XM_017029063.1
XP_016884552.1
5
894
XM_017029064.1
XP_016884553.1
4
777
GenBank mRNA
AB051454.1
-
5
897
AK054717.1
-
3
418
AK057648.1
-
5
823
AK290211.1
-
5
914
AL365512.1
-
3
515
AL713795.1
-
5
943
AY043416.1
-
4
514
BC028625.1
-
5
899
BC050285.1
-
5
876
BC065030.1
-
4
716
6148557
CR456392.1
-
4
415
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting
lysosome organization
blood coagulation
hemostasis
melanocyte differentiation
positive regulation of eye pigmentation
protein stabilization
positive regulation of protein targeting to mitochondrion
protein binding
protein homodimerization activity
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0