Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167738.1 | NP_001161210.1 | 23 | 4020 | ||
NM_020443.4 | NP_065176.3 | 26 | 5360 | |||
XM_006711609.2 | XP_006711672.2 | 24 | 3995 | |||
XM_006711610.2 | XP_006711673.2 | 23 | 3971 | |||
XM_006711611.1 | XP_006711674.1 | 24 | 4111 | |||
XM_011510097.1 | XP_011508399.1 | 25 | 5030 | |||
XM_011510098.1 | XP_011508400.1 | 25 | 5015 | |||
XM_011510099.1 | XP_011508401.1 | 24 | 5006 | |||
XM_011510100.1 | XP_011508402.1 | 25 | 4868 | |||
XM_011510101.1 | XP_011508403.1 | 24 | 4844 | |||
XM_011510102.1 | XP_011508404.1 | 23 | 4835 | |||
XM_017002751.1 | XP_016858240.1 | 24 | 4859 | |||
XM_017002752.1 | XP_016858241.1 | 22 | 3962 | |||
GenBank mRNA | AB032977.3 | - | 26 | 4693 | ||
AJ488101.1 | - | 26 | 4891 | |||
AK022622.1 | - | 4 | 625 | |||
AK024265.1 | - | 11 | 1525 | |||
AL133664.1 | - | 1 | 462 | |||
AY043013.1 | - | 26 | 5360 | |||
BC007523.1 | - | 12 | 1642 | |||
BX648460.1 | - | 23 | 3987 |