Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG16L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318766.1 | NP_001305695.1 | 10 | 1182 | ||
NM_033388.1 | NP_203746.1 | 10 | 1062 | |||
XM_005274376.4 | XP_005274433.1 | 10 | 1061 | |||
XM_006718732.2 | XP_006718795.1 | 9 | 998 | |||
XM_011545332.1 | XP_011543634.1 | 9 | 947 | |||
XM_011545333.1 | XP_011543635.1 | 10 | 1510 | |||
XM_011545334.1 | XP_011543636.1 | 10 | 923 | |||
GenBank mRNA | AK024423.1 | - | 3 | 2799 | ||
AK091151.1 | - | 9 | 2650 | |||
AK093017.1 | - | 10 | 1035 | |||
AL832974.1 | - | 2 | 673 | |||
BC036713.2 | - | 8 | 791 | |||
BC137489.1 | - | 10 | 1160 | 9021489 | ||
BC137490.1 | - | 10 | 1160 | 9021490 | ||
BC142718.1 | - | 8 | 2758 | |||
BC146660.1 | - | 8 | 2758 |