Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033123.3 | NP_149114.2 | 10, 11 | 1437 | ||
NR_073075.1 | 10, 11 | 1440 | ||||
XM_005253519.2 | XP_005253576.1 | 9, 10 | 1553 | |||
XM_011520883.1 | XP_011519185.1 | 7, 8 | 843 | |||
XM_017020176.1 | XP_016875665.1 | 9, 10 | 1553 | |||
XM_017020177.1 | XP_016875666.1 | 10, 11 | 1430 | |||
XM_017020178.1 | XP_016875667.1 | 8, 9 | 1403 | |||
XM_017020179.1 | XP_016875668.1 | 9, 10 | 1548 | |||
XM_017020180.1 | XP_016875669.1 | 9, 10 | 1442 | |||
XM_017020181.1 | XP_016875670.1 | 9, 10 | 1507 | |||
XM_017020182.1 | XP_016875671.1 | 5, 6 | 1206 | |||
XM_017020183.1 | XP_016875672.1 | 6, 7 | 970 | |||
XM_017020184.1 | XP_016875673.1 | 6, 7 | 898 | |||
XM_017020185.1 | XP_016875674.1 | 5, 6 | 783 | |||
XM_017020186.1 | XP_016875675.1 | 7, 8 | 805 | |||
GenBank mRNA | AF532185.1 | - | 10, 11 | 1362 | ||
AK097555.1 | - | 6, 7 | 785 | |||
AK097725.1 | - | 6, 7 | 903 | |||
AK292279.1 | - | 10, 11 | 1435 | |||
AK314516.1 | - | 10, 11 | 1394 | |||
AY035866.1 | - | 10, 11 | 1399 | |||
BC125067.1 | - | 6, 7 | 690 | 40117397 |