Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033229.2 | NP_150232.2 | 1 | 760 | ||
XM_011514988.2 | XP_011513290.1 | 1 | 281 | |||
GenBank mRNA | AF220132.1 | - | 1 | 281 | ||
AF220133.1 | - | 1 | 281 | |||
AK290280.1 | - | 1 | 466 | |||
AK291853.1 | - | 1 | 418 | |||
BC038585.1 | - | 1 | 361 | 5758860 | ||
U34249.1 | - | 1 | 774 |