Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM116 SIRNA Assays ›
Gene Symbol:
TMEM116
Gene Name:
transmembrane protein 116
Gene Aliases:
-
Chromosome Location:
Chr.12: 111894936 - 112013219 on Build GRCh38
UniGene ID:
Hs.506815
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s40199
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM116
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001193453.1
NP_001180382.1
6
545
NM_001193531.1
NP_001180460.1
7
650
NM_001294314.1
NP_001281243.1
9
867
NM_138341.2
NP_612350.1
6
586
NR_122119.1
6
586
XM_006719678.3
XP_006719741.1
7
552
XM_011538946.1
XP_011537248.1
7
504
XM_011538947.1
XP_011537249.1
6
399
XM_011538948.2
XP_011537250.1
7
504
XM_011538949.2
XP_011537251.1
7
455
XM_011538950.1
XP_011537252.1
7
504
XM_011538951.2
XP_011537253.1
6
423
XM_011538953.1
XP_011537255.1
9
840
XM_011538955.1
XP_011537257.1
6
447
XM_011538957.1
XP_011537259.1
8
761
XM_011538958.1
XP_011537260.1
7
656
XM_017020187.1
XP_016875676.1
7
504
XM_017020188.1
XP_016875677.1
8
724
XM_017020189.1
XP_016875678.1
7
612
XM_017020190.1
XP_016875679.1
8
669
XM_017020191.1
XP_016875680.1
6
533
XM_017020192.1
XP_016875681.1
8
2174
XM_017020193.1
XP_016875682.1
4
263
XM_017020194.1
XP_016875683.1
5
428
GenBank mRNA
AK074648.1
-
9
779
AL519857.3
-
6
502
AL833501.1
-
7
571
BC000282.1
-
6
465
BC022360.1
-
2
74
BC048796.1
-
7
542
5193616
BC070135.1
-
6
492
3839474
BI828772.1
-
7
650
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline