Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130466.3 | NP_569733.2 | 17, 18 | 2470 | ||
NM_183415.2 | NP_904324.1 | 17, 18 | 2125 | |||
XM_005253987.2 | XP_005254044.1 | 17, 18 | 2350 | |||
XM_006719681.3 | XP_006719744.1 | 17, 18 | 2683 | |||
XM_006719682.2 | XP_006719745.1 | 17, 18 | 2683 | |||
XM_011538959.2 | XP_011537261.1 | 16, 17 | 2232 | |||
XM_011538961.1 | XP_011537263.1 | 17, 18 | 2683 | |||
XM_017020195.1 | XP_016875684.1 | 17, 18 | 2893 | |||
GenBank mRNA | AF130072.1 | - | 15, 16 | 1862 | ||
AF251046.2 | - | 17, 18 | 2440 | |||
AK127227.1 | - | 2, 3 | 1257 | |||
AK307385.1 | - | 16, 17 | 2004 | |||
AL096740.1 | - | 17, 18 | 2439 | |||
BC020348.1 | - | 2, 3 | 164 | |||
BC032301.1 | - | 17, 18 | 2103 | |||
BC141880.1 | - | 17, 18 | 2348 | 40124513 | ||
BC144684.1 | - | 17, 18 | 2446 | |||
BC150637.1 | - | 17, 18 | 2101 | 9057007 | ||
BX537403.1 | - | 17, 18 | 2115 |