Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142469.1 | NP_001135941.1 | 4 | 327 | ||
NM_033201.2 | NP_149978.1 | 4 | 508 | |||
XM_011522731.2 | XP_011521033.1 | 4 | 487 | |||
XM_017023824.1 | XP_016879313.1 | 5 | 634 | |||
GenBank mRNA | AK057180.1 | - | 4 | 508 | ||
AK293431.1 | - | 4 | 414 | |||
BC008967.2 | - | 4 | 438 | 3010666 | ||
BC023603.2 | - | 4 | 327 | 4330351 |