Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142469.1 | NP_001135941.1 | 2, 3 | 237 | ||
NM_033201.2 | NP_149978.1 | 2, 3 | 418 | |||
XM_011522731.2 | XP_011521033.1 | 2, 3 | 397 | |||
GenBank mRNA | AK057180.1 | - | 2, 3 | 418 | ||
AK293431.1 | - | 2, 3 | 324 | |||
BC008967.2 | - | 2, 3 | 348 | 3010666 | ||
BC023603.2 | - | 2, 3 | 237 | 4330351 |