Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173462.3 | NP_775733.3 | 7, 8 | 694 | ||
GenBank mRNA | BC144315.1 | - | 6, 7 | 612 | ||
BX470414.1 | - | 3, 4 | 353 |