Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001357.4 | NP_001348.2 | 8 | 948 | ||
NR_033302.1 | 9 | 1085 | ||||
GenBank mRNA | AK226102.1 | - | 9 | 1066 | ||
AK308165.1 | - | 8 | 902 | |||
BC008773.1 | - | 8 | 865 | |||
BC014246.2 | - | 8 | 883 | |||
BC025245.1 | - | 8 | 870 | |||
BC137136.1 | - | 8 | 895 | 9021133 | ||
BC144494.1 | - | 8 | 895 | |||
Y10658.1 | - | 7 | 773 |